Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNMT Antibody (1C11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP197942AF
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197942AF 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197942AF Supplier R&D Systems Supplier No. NBP197942AF405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

PNMT Monoclonal specifically detects PNMT in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen PNMT
Applications ELISA, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1C11
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase
Gene Symbols PNMT
Host Species Mouse
Immunogen Recombinant human PNMT (1-282aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5409
Test Specificity Anti-human PNMT mAb, clone 1C11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PNMT protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.