Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544563 Supplier Invitrogen™ Supplier No. PA544563
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MAVAVRTLQE QLEKAKESLK NVDENIRKLT GRDPNDVRPI QARLLALSGP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Transcriptional activator binding to the E-box 1 core sequence of the E-cadherin promoter gene; the core-binding sequence is 5'CAGGTG-3'. Capable of reversing CTBP1-mediated transcription repression. Auxiliary component of the splicing-dependent multiprotein exon junction complex (EJC) deposited at splice junction on mRNAs. The EJC is a dynamic structure consisting of core proteins and several peripheral nuclear and cytoplasmic associated factors that join the complex only transiently either during EJC assembly or during subsequent mRNA metabolism. Participates in the regulation of alternative pre-mRNA splicing. Associates to spliced mRNA within 60 nt upstream of the 5'-splice sites. Component of the PSAP complex which binds RNA in a sequence-independent manner and is proposed to be recruited to the EJC prior to or during the splicing process and to regulate specific excision of introns in specific transcription subsets. Involved in the establishment and maintenance of epithelia cell-cell adhesion. Potential tumor suppressor for renal cell carcinoma. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNN
Gene Accession No. Q9H307
Gene Alias 140 kDa nuclear and cell adhesion-related phosphoprotein; AU045199; D12Ertd512e; Desmosome-associated protein; domain-rich serine protein; DRS; DRS protein; DRSP; fb24d03; Melanoma metastasis clone A protein; memA; neutrophil protein; nuclear protein SDK3; Pinin; pinin, desmosome associated protein; PNN; PNN protein; SDK3; SR-like protein; wu:fb24d03
Gene Symbols PNN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PNN (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5411
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.