Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541923 Supplier Invitrogen™ Supplier No. PA541923
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CGGSLALMDS GVPISSAVAG VAIGLVTKTD PEKGEIEDYR LLTDILGIED Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 93%; Rat: 86%.

PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPase
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PNPT1
Gene Accession No. Q8TCS8
Gene Alias 1200003F12Rik; 3'-5' RNA exonuclease OLD35; COXPD13; DFNB70; DKFZp762K1914; old 35; Old35; old-35; PNPase; PNPase 1; PNPase old-35; Pnpt1; Pnptl1; polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; polyribonucleotide nucleotidyltransferase 1; polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Symbols PNPT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PNPT1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 87178
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.