Learn More
Abcam oNPG, beta-galactosidase substrate
Supplier:  Abcam AB1463925G
Encompass_Preferred
MW 301.25 g/mol. β-galactosidase substrate for colorimetric and EIA applications. Hydrolysed by β-galactosidase to yield galactose and ortho-Nitrophenol, which is yellow in color.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4660
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.