Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPO Monoclonal Antibody (OTI1H9)
GREENER_CHOICE

Catalog No. PIMA525659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525659 Supplier Invitrogen™ Supplier No. MA525659
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PNPO Monoclonal Antibody for Western Blot, IHC (P)

The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H9
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PNPO
Gene Accession No. Q9NVS9
Gene Alias AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase
Gene Symbols PNPO
Host Species Mouse
Immunogen Full length human recombinant protein of PNPO produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.