Learn More
Invitrogen™ PNPO Polyclonal Antibody

Description
Immunogen sequence: MTCWLRGVTA TFGRPAEWPG YLSHLCGRSA AMDLGPMRKS YRGDREAFEE THLTSLDPVK QFAAWFEEAV QCPDIGEANA MCLATCTRDG KPSARMLLLK GFGKDGFRFF TNFESRKGKE LDSNPFASLV FYWEPLNRQV RVEGPVKKLP EEEAECYFHS RPKSSQIGAV VSHQSSVIPD REYLRKKNEE LEQLYQDQEV PKPKSWGGYV LYPQVMEFWQ GQTNRLHDRI VFRRGLPTGD SPLGPMTHRG EEDWLYERLA P.
Specifications
Specifications
| Antigen | PNPO |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.49 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PNPO |
| Gene Accession No. | O88794, Q91XF0, Q9NVS9 |
| Gene Alias | AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase |
| Gene Symbols | PNPO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.