Learn More
Bioassay Systems pNPP Phosphatase Assay Kit. For quantitative determination of phosphatase activity.
SDP
Catalog No. 50489297 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of phosphatase enzyme activity. Key Features: High sensitivity and wide linear range. The detection limit is generally 3 ng phosphatase or below. Homogeneous and simple procedure. No wash or reagent transfer steps are involved. The assay can be completed within 30 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Protein phosphatases. Species: All. Procedure: Assay takes 30 min. Kit size: 500 tests. Detection limit: 3 ng.
Catalog No. 50-489-297 Supplier Bioassay Systems Supplier No. POPN500
For quantitative determination of phosphatase enzyme activity. Key Features: High sensitivity and wide linear range. The detection limit is generally 3 ng phosphatase or below. Homogeneous and simple procedure. No wash or reagent transfer steps are involved. The assay can be completed within 30 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Protein phosphatases. Species: All. Procedure: Assay takes 30 min. Kit size: 500 tests. Detection limit: 3 ng.