Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112659
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112659 Supplier Invitrogen™ Supplier No. PA5112659
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PNRC2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

PNRC2 gene ontology annotations related to this gene include nuclear-transcribed mRNA catabolic process, nonsense-mediated decay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNRC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PNRC2
Gene Accession No. Q9NPJ4
Gene Alias 0610011E17Rik; D4Bwg0593e; HSPC208; Pnrc2; proline rich nuclear receptor coactivator 2; proline-rich nuclear receptor coactivator 2; proline-rich nuclear receptor coactivator-like protein
Gene Symbols PNRC2
Host Species Rabbit
Immunogen Recombinant Human Proline-rich nuclear receptor coactivator 2 protein (1-139AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55629
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.