Learn More
Selleck Chemical LLC PNU-74654 10mg 113906-27-7
SDP
Supplier:  Selleck Chemical LLC S842910MG
Encompass_Preferred
PNU-75654 disrupts the Wnt signaling pathway via inhibition of the interaction (KD50=450 nM) between ?-catenin and Tcf4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6052
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.