Learn More
Roche Diagnostics POC Chemstrip Urine Test Strips
Catalog No. 22045620 Shop All Roche Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Leukocyte, Nitrite, Protein
Glucose, Protein
Leukocyte, Nitrite
Type:
Chemstrip 10 MD
Chemstrip 10 SG
Chemstrip 2 GP
Chemstrip 2 LN
Chemstrip 5 OB
Chemstrip 7
Chemstrip 9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Type
22-045-620 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein Chemstrip 7
BC417145 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Chemstrip 10 SG
22-045-621 Blood, Glucose, Leukocyte, Nitrite, Protein Chemstrip 5 OB
BC417109 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen Chemstrip 9
BC200743 Glucose, Protein Chemstrip 2 GP
22-208-840 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Chemstrip 10 MD
BC417152 Leukocyte, Nitrite Chemstrip 2 LN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-045-620 Supplier Roche Diagnostics Supplier No. 11008552160
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accuracy, performance and testing simplicity
  • Avoid potentially costly consequences of missed diagnoses — vitamin C interference is virtually eliminated on the glucose pad1, 2
  • Avoid false negatives — both acetoacetic acid and acetone are detected on the ketone reagent pad3
  • Simplify and streamline reading — all reagent pads are read at one time, between one and two minutes after dipping3
  • Promote safe and hygienic testing practice — fingers stay clear of specimen
  • Generate confidence in test result accuracy — Chemstrip construction allows uniform liquid penetration and prevents runover onto adjacent pads
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
Detectable Analytes Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Type Chemstrip 7
For Use With (Application) Urinalysis
Quantity 100 Pack