Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PocketTips™ Pipette Tips

Catalog No. 14387371
Change view
Click to view available options
Volume (Metric):
100 nL
250 nL
50 nL
500 nL
For Use With (Equipment):
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
Hamilton MICROLAB STAR
Molecular Devices FLIPR 384-well Head
Packard 384-well, PerkinElmer
Packard 96-well, PerkinElmer
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
14-387-371 500 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
14-387-363 250 nL Molecular Devices FLIPR 384-well Head
14-387-364 50 nL Molecular Devices FLIPR 384-well Head
14-387-365 100 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
14-387-366 250 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
14-387-367 50 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
14-387-368 100 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
14-387-370 50 nL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
14-387-372 100 nL Packard 384-well, PerkinElmer
14-387-373 50 nL Packard 384-well, PerkinElmer
14-387-374 100 nL Packard 96-well, PerkinElmer
14-387-375 250 nL Packard 96-well, PerkinElmer
14-387-376 100 nL Hamilton MICROLAB STAR
14-387-377 250 nL Hamilton MICROLAB STAR
14-387-378 50 nL Hamilton MICROLAB STAR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 14-387-371 Supplier Thermo Scientific™ Supplier No. FX96P20500

Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.

Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.

Features:

  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing
  • Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
  • Color: Clear

Recommended for:

Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 500 nL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
Product Line PocketTip

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.