Learn More
EMD MILLIPORE 3P BIOPHARM HC Pro Pod Depth Filter, X0SP media series, 1.1 m2 surface area
Supplier:  EMD MILLIPORE 3P BIOPHARM MX0SP10FS1
Encompass
Secondary clarification following primary clarification or primary clarification depth filter.
Catalog No. NC1638246
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.