Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Podocalyxin Monoclonal Antibody (B34D1.3), PE, eBioscience™
GREENER_CHOICE

Catalog No. 50245533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-533 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-533 Supplier Invitrogen™ Supplier No. 12887342
  • $342.00
    $256.65 / Each of 1

    Save $85.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The monoclonal antibody B34D1.3 recognizes human podocalyxin, a protein expressed on undifferentiated human embryonic stem cells (ES), embryonic carcinoma cells (EC), and embryonic germ cells (EG). Podocalxyin is a heavily glycosylated cell adhesion protein with homology to CD34 and endoglycan. It was first identified on kidney podocytes, and then found to be upregulated in a variety of human cancers such as breast and prostate cancer. Like other stem cell-specific markers, the epitope recognized by the B34D1.3 antibody is lost upon cell differentiation. The B34D1.3 antibody recognizes the ectodomain of podocalyxin, which is a different epitope from the Tra-1-60 and Tra-1-81 monoclonal antibodies. While Tra1-60 has been found to react with a sialidase-sensitive epitope, the epitope recognized by Tra1-81 has not been fully defined. Applications Reported: This B34D1.3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B34D1.3 antibody has been pre-titrated and tested by flow cytometric analysis of the NTERA cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podocalyxin
Applications Flow Cytometry
Classification Monoclonal
Clone B34D1.3
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene PODXL
Gene Accession No. O00592
Gene Alias AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl
Gene Symbols PODXL
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.