Learn More
Medchemexpress LLC Podophyllotoxone | 477-49-6 | 99.9% | 412.39 g·mol⁻1 | C22H20O8 | 10 MG

Supplier: Medchemexpress LLC HYN241510MG
Podophyllotoxone is a lignan-class tubulin inhibitor isolated from the roots of Dysosma versipellis. It exhibits antiproliferative activity by inhibiting tubulin polymerization and is supplied as a research-grade small-molecule reagent for in vitro studies of microtubule dynamics and antimitotic effects.
- High purity: 99.94%.
- Molecular formula: C22H20O8; molecular weight: 412.39 g·mol⁻1.
- Reported bioactivity: inhibits PC3 and DU145 cell growth (IC50 = 14.7 μM and 20.6 μM).
- Appearance: solid; storage at 4°C, protected from moisture and light.
- Intended for research use in studies of microtubule/tubulin function.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.