Learn More
Medchemexpress LLC HY-N1416 5mg Medchemexpress, Pogostone CAS:23800-56-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14165MG
Encompass_Preferred
Medchemexpress, HY-N1416 5mg Pogostone CAS:23800-56-8 Pogostone is isolated from patchouli with anti-bacterial and anti-cancer activities. Pogostone inhibits both gram negative and gram positive bacteria, also show inhibitory effect on corynebacterium xerosis with a MIC value of 0.098 µg/ml . Pogostone induces cell apoptosis and autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.