Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Acid Phosphatase Reagents

Catalog No. 23666059 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
196 Tests/Kit
For Use With (Equipment):
Hitachi Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Equipment)
23-666-059 196 Tests/Kit Hitachi Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-059 Supplier Pointe Scientific Supplier No. A7503100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Set 1 includes Acid Phosphatase Reagent (10 x 10mL), L-Tartrate Reagent (5mL), and Acetate Buffer (15mL). Reagent Kit HA703-60 is used with Hitachi 704, 717.

Specifications

Certifications/Compliance CE Marked
Type Acid Phosphatase Reagent Kit
Formulation Acid Phosphatase reagent: alpha-naphthylphosphate 3mM, Fast Red TR 1mM, Citric Acid 20mM, Sodium Citrate 60mM pH 5.3 ± 0.1, L-Tartrate reagent: Sodium L Tartrate 2M, Citric Acid 70mM, Sodium Citrate 10mM pH 5.3 ± 0.1, A
For Use With (Application) Quantitative determination of Acid Phosphatase in Serum
For Use With (Equipment) Hitachi Analyzer
Includes 10 x 10 mL Reagent, 1 x 5 mL L-Tartrate, 1 x 15 mL Acetate Buffer
Quantity 196 Tests/Kit
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.