Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Alkaline Phosphatase Reagents

Catalog No. 23666079 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 100mL (R1); 4 x 23mL (R2)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-079 4 x 100mL (R1); 4 x 23mL (R2)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-079 Supplier Pointe Scientific Supplier No. HA916492

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For quantitative determination of Alkaline Phosphatase in human serum

HA716-246 is for Hitachi 704, 717. HA916-492 is for Hitachi 911, 912, 914.

  • Format: Dry Powder
  • Wavelength: 405nm
  • Linearity: 800 U/L
  • Expected Values: Adults 35-123 U/L, Children have a higher normal value
  • Reconstituted Stability: 7 days RT, 60 days 2°-8°C

Specifications

Certifications/Compliance CE Marked
Type Alkaline Phosphatase Reagent Kit
Formulation R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives
For Use With (Application) Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
pH 10.45
Quantity 4 x 100mL (R1); 4 x 23mL (R2)
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.