Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ALT Reagents for Hitachi Analyzers

Catalog No. 23666099 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 253 R2mL
5 x 500 R1mL
5 x 50mL R1, 1 x 50mL R2
Formulation:
Ketoglutaric Acid 0.2%, Sodium Azide 0.095%
Ketoglutaric Acid 0.2%, Sodium Azide 1%
R1 and R2 contains: L-alanine 500mM, alpha-ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM, pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers
For Use With (Equipment):
Hitachi 704 analyzers
Hitachi 747 analyzer
Type:
ALT R1 Kit
ALT R2 Kit
ALT Reagent Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation For Use With (Equipment) Type
23-666-099 5 x 253 R2mL Ketoglutaric Acid 0.2%, Sodium Azide 1% Hitachi 747 analyzer ALT R2 Kit
23-666-098 5 x 500 R1mL Ketoglutaric Acid 0.2%, Sodium Azide 0.095% Hitachi 747 analyzer ALT R1 Kit
23-666-095 5 x 50mL R1, 1 x 50mL R2 R1 and R2 contains: L-alanine 500mM, alpha-ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM, pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers Hitachi 704 analyzers ALT Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-099 Supplier Pointe Scientific Supplier No. HA426R2

missing translation for 'validMainframeMsg'

For quantitative determination of alanine aminotransferase in serum on Hitachi analyzers

HA726-305 is for Hitachi 704. HA926-610 is for Hitachi 911, 912, 914. HA426-R1 and HA426-R2 for Hitachi 747.

  • Method: Modified IFCC, Kinetic
  • Format: Liquid
  • Linearity: 0-500 U/L
  • Expected Values: 4-36 U/L
  • Storage Temperature: 2° to 8°C
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type ALT R2 Kit
Formulation Ketoglutaric Acid 0.2%, Sodium Azide 1%
For Use With (Application) Quantitative determination of Alanine Aminotransferase in Serum
For Use With (Equipment) Hitachi 747 analyzer
Quantity 5 x 253 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.