Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ALT (SGPT) Liquid Reagents

Catalog No. 23666089 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120mL R1, 1 x 30mL R2
1 x 500mL R1, 1 x 125mL R2
3 x 120mL R1, 1 x 90mL R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-089 1 x 500mL R1, 1 x 125mL R2
23-666-087 1 x 120mL R1, 1 x 30mL R2
23-666-088 3 x 120mL R1, 1 x 90mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-089 Supplier Pointe Scientific Supplier No. A7526625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For quantitative determination of Alanine Aminotransferase in serum

Reagent Set 1 includes 1 x 120mL substrate/enzyme reagent and 1 x 30mL coenzyme reagent.
Reagent Set 2 includes 3 x 120mL substrate/enzyme reagent and 1 x 90mL coenzyme reagent.
Reagent Set 3 includes 1 x 500mL substrate/enzyme reagent and 1 x 125mL coenzyme reagent.

  • Method: Modified IFCC, Kinetic
  • Format: Liquid
  • Wavelength: 340nm
  • Linearity: 0-500 U/L
  • Expected Values: 4-36 U/L
  • Storage Temperature: 2° to 8°C
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type Alanine Aminotransferase Reagent
Formulation L-alanine 500mM, alpha-Ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers
For Use With (Application) Quantitative determination of Alanine Aminotransferase in Serum
Quantity 1 x 500mL R1, 1 x 125mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.