Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Ammonia Reagents

Catalog No. 23666103 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
10 x 6.5mL Substrate, 10 x 2mL Enzyme
4 x 6.5mL Substrate, 4 x 2mL Enzyme
Formulation:
Ammonia Enzyme/R2: Buffer 100mM/L, EDTA 2mM/L, Adenosine Diphosphate 0.5mM/L, GLDH 400 Kμ/L, fillers and Stabilizers, pH 7.8 ± 0.1
Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1
Includes:
Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
Reagent set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of Enzyme reagent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation Includes
23-666-103 4 x 6.5mL Substrate, 4 x 2mL Enzyme Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1 Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
23-666-102 10 x 6.5mL Substrate, 10 x 2mL Enzyme Ammonia Enzyme/R2: Buffer 100mM/L, EDTA 2mM/L, Adenosine Diphosphate 0.5mM/L, GLDH 400 Kμ/L, fillers and Stabilizers, pH 7.8 ± 0.1 Reagent set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of Enzyme reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-103 Supplier Pointe Scientific Supplier No. A755334
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of enzyme reagent.
Reagent Set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of enzyme reagent.

Specifications

Certifications/Compliance CE Marked
Type Ammonia Reagent
Formulation Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1
For Use With (Application) Enzymatic measurement of Ammonia in Plasma, in-vitro diagnostic use only
Includes Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
Quantity 4 x 6.5mL Substrate, 4 x 2mL Enzyme
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.