Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Amylase-EPS Reagents

Catalog No. 23666109 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
3 x 120 R2mL
3 x 270 R1mL
6 x 22mL R1, 3 x 10mL R2
Formulation:
Buffer 50mM pH 7.2 ±0.1, NaCl >50mM, MgCl2 10mM, alpha-Glucosidase >2KU/L, EPS-G7 >1mM
Sodium Azide 0.095%
For Use With (Equipment):
Hitachi 704 analyzers
Hitachi 747 analyzers
Type:
Amylase-EPS R1
Amylase-EPS R2
Amylase-EPS Reagent Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation For Use With (Equipment) Type
23-666-109 3 x 120 R2mL Sodium Azide 0.095% Hitachi 747 analyzers Amylase-EPS R2
23-666-108 3 x 270 R1mL Sodium Azide 0.095% Hitachi 747 analyzers Amylase-EPS R1
23-666-107 6 x 22mL R1, 3 x 10mL R2 Buffer 50mM pH 7.2 ±0.1, NaCl >50mM, MgCl2 10mM, alpha-Glucosidase >2KU/L, EPS-G7 >1mM Hitachi 704 analyzers Amylase-EPS Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-109 Supplier Pointe Scientific Supplier No. HA465R2

missing translation for 'validMainframeMsg'

HA965-324 is for Hitachi 911, 912, 914. HA765-162 is for Hitachi 704. HA465-R1 and HA465-R2 are for Hitachi 747.

Specifications

Certifications/Compliance CE Marked
Type Amylase-EPS R2
Formulation Sodium Azide 0.095%
For Use With (Application) Quantitative kinetic determination of a-amylase activity in Human Serum
For Use With (Equipment) Hitachi 747 analyzers
Quantity 3 x 120 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.