Learn More
Description
HA761-305 is for Hitachi 704. HA961-610 is for Hitachi 911, 912, 914. HA461-R1 and HA461-R2 are for Hitachi 747.
- Modified IFCC, Kinetic
- Format: Liquid
- Linearity: 0-500 U/L
- Expected Values: 5-34 U/L
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | AST R1 |
| Formulation | Ketoglutaric Acid 0.2%, Sodium Azide 0.095% |
| For Use With (Application) | Quantitative determination of Aspartate Aminotransferase in Human Serum |
| For Use With (Equipment) | Hitachi 747 Analyzers |
| Quantity | 5 x 500 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.