Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific AST Reagents
Click to view available options
Quantity:
5 x 100 mL R1, 2 x 50 mL R2
5 x 253 mL
5 x 50 mL R1, 1 x 50 mL R2
5 x 500 mL
Type:
AST R1
AST R2
AST Reagent Kit
For Use With (Equipment):
Hitachi 704 Analyzers
Hitachi 717 Analyzers
Hitachi 747 Analyzers
Description
HA761-305 is for Hitachi 704. HA961-610 is for Hitachi 911, 912, 914. HA461-R1 and HA461-R2 are for Hitachi 747.
- Modified IFCC, Kinetic
- Format: Liquid
- Linearity: 0-500 U/L
- Expected Values: 5-34 U/L
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | AST R2 |
Formulation | Ketoglutaric Acid 0.2%, Sodium Azide 1% |
For Use With (Application) | Quantitative determination of Aspartate Aminotransferase in Human Serum |
For Use With (Equipment) | Hitachi 747 Analyzers |
Quantity | 5 x 253 mL |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction