Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Auto LDL Cholesterol Liquid Reagents

Catalog No. 23666143 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 240mL R1, 1 x 80mL R2
1 x 30mL R1, 1 x 10mL R2
1 x 60mL R1, 1 x 20mL R2
Type:
AutoLDL™ Cholesterol Reagent
AutoLDL™ Cholesterol Reagent Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-143 1 x 60mL R1, 1 x 20mL R2 AutoLDL™ Cholesterol Reagent
23-666-141 1 x 240mL R1, 1 x 80mL R2 AutoLDL™ Cholesterol Reagent
23-666-142 1 x 30mL R1, 1 x 10mL R2 AutoLDL™ Cholesterol Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-143 Supplier Pointe Scientific Supplier No. L757480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For direct quantitative determination of low density lipoprotein cholesterol (LDL-C) in human serum or plasma

Requires autoHDL/LDL Calibrator (23-666-140).

  • Method: Direct/Automated
  • Format: Liquid
  • Wavelength: 546/660nm
  • Linearity: 0-700mg/dL
  • Expected Values: >130mg/dL
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type AutoLDL™ Cholesterol Reagent
Formulation R1 contains: MES Buffer pH 6.3, Detergent 1, Cholesterol Esterase, Cholesterol Oxidase, Peroxidase, 4-aminoantipyrine, Ascorbic Acid Oxidase, Preservative, R2 contains: MES Buffer pH 6.3, Detergent 2, N,N-bis (4-sulfhobutyl)-m Toluidine-disodium (DSBmT),
For Use With (Application) Quantitative determination of Low Density Lipoprotein Cholesterol in Human Serum or Plasma
Quantity 1 x 60mL R1, 1 x 20mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.