Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific autoHDL Cholesterol Reagents

Catalog No. 23666135 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
3 x 150 R2mL
5 x 250 R1mL
6 x 30mL R1, 3 x 20mL R2
Type:
AutoHDL Cholesterol Reagent Kit
AutoHDL™ Cholesterol R1
AutoHDL™ Cholesterol R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-135 3 x 150 R2mL AutoHDL™ Cholesterol R2
23-666-134 5 x 250 R1mL AutoHDL™ Cholesterol R1
23-666-133 6 x 30mL R1, 3 x 20mL R2 AutoHDL Cholesterol Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-135 Supplier Pointe Scientific Supplier No. HH445R2

missing translation for 'validMainframeMsg'

Requires autoHDL/LDL Calibrator (23-666-140). HH945-240 is for Hitachi 911, 912, 914. HH445-R1 and HH4450R2 are for Hitachi 747.

Specifications

Certifications/Compliance CE Marked
Type AutoHDL™ Cholesterol R2
Formulation R2: POD >15000U/L, PEG-CO >5000U/L, PEG-CE >800U/L, 4-aminoantipyrene 0.5g/L, Buffer pH 7 ± 0.1, Surfactant, preservative
For Use With (Application) Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma
Quantity 3 x 150 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.