Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific BUN Reagents

Catalog No. 23666161 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 300 R2mL
5 x 600 R1mL
8 x 50mL R1, 2 x 50mL R2
Type:
BUN R1
BUN R2
BUN Reagent Kit
For Use With (Equipment):
Hitachi 704 analyzers
Hitachi 747 analyzers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type For Use With (Equipment)
23-666-161 5 x 300 R2mL BUN R2 Hitachi 747 analyzers
23-666-160 5 x 600 R1mL BUN R1 Hitachi 747 analyzers
23-666-154 8 x 50mL R1, 2 x 50mL R2 BUN Reagent Kit Hitachi 704 analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-161 Supplier Pointe Scientific Supplier No. HB452R2

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance CE Marked
Type BUN R2
Formulation Ketoglutaric Acid 0.2%, Sodium Azide 1%
For Use With (Application) Quantitative determination of Urea Nitrogen in Serum
For Use With (Equipment) Hitachi 747 analyzers
Quantity 5 x 300 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.