Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Urea Nitrogen (BUN) UV Reagent Kit |
| Formulation | NADH 0.3mM, Urease 1,500U/L, Glutamate Dehydrogenase >1500U/L, alpha-Ketoglutarate 4mM, pH 8.2 ± 0.1 Buffer, activators and non-reactive Stabilizers |
| For Use With (Application) | Quantitative determination of Urea Nitrogen in Serum, in-vitro diagnostic use only |
| Quantity | 300 mL |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.