Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Calcium Liquid Reagents

Catalog No. 23666168 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
2 x 120mL Color, 2 x 120mL Buffer
2 x 30mL Color, 2 x 30mL Buffer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-168 2 x 120mL Color, 2 x 120mL Buffer
23-666-169 2 x 30mL Color, 2 x 30mL Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-168 Supplier Pointe Scientific Supplier No. C7503480
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Color Reagent: o-cresolphthalein complexone 0.11mM, 8-hydroxyquinoline 17.0mM, surfactant
  • Buffer Reagent: 2-Amino-2-Methyl-1-Propanol 976mM, Potassium Cyanide 2.0mM

Quantify calcium levels in serum

  • Method: Cresolphthalein Complexone (CPC), Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 570nm
  • Linearity: 20mg/dL
  • Expected Values: 8.5 to 10.4mg/dL
  • Separate color and buffer reagents

For quantitative determination of calcium in serum

Specifications

Certifications/Compliance CE Marked
Type Calcium Reagent Kit
For Use With (Application) Quantitative determination of Calcium in Serum
Includes Color reagent : o-Cresolphthalein Complexone 0.11mM, 8-Hydroxyquinoline 17.0mM, Surfactant, Buffer reagent : 2-Amino-2-Methyl-1-Propanol 976mM, 2mM Potassium Cyanide
Quantity 2 x 120mL Color, 2 x 120mL Buffer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.