Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Calcium Reagents for Hitachi™ Analyzers

Catalog No. 23666181 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 300 mL R2
5 x 600 mL R1
7 x 100 mL R1, 3 x 100 mL R2
7 x 50 mL R1, 3 x 50 mL R2
For Use With (Equipment):
Hitachi 704, 902 Analyzers
Hitachi 717, 911, 912, 914 Analyzers
Hitachi 747 Analyzers
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Equipment)
23-666-181 5 x 300 mL R2 Hitachi 747 Analyzers
23-666-180 5 x 600 mL R1 Hitachi 747 Analyzers
23-666-179 7 x 50 mL R1, 3 x 50 mL R2 Hitachi 704, 902 Analyzers
23-666-178 7 x 100 mL R1, 3 x 100 mL R2 Hitachi 717, 911, 912, 914 Analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-666-181 Supplier Pointe Scientific Supplier No. HC402R2

missing translation for 'validMainframeMsg'

Includes

  • Reagent 1: 2-Amino-2-Methyl-1-Propanol, pH 10.6±0.1
  • Reagent 2: o-CPC 0.3mM, surfactant

Quantify calcium levels easily with these instrument-specific reagents

  • Method: Cresolphthalein Complexone (CPC)
  • Format: Liquid (ready to use)
  • Linearity: 20mg/dL
  • Expected Values: 8.5 to 10.5mg/dL

Applications:

For quantitative determination of calcium in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Calcium Reagent Kit
Formulation o-CPC 0.3mM, Surfactant
For Use With (Application) Quantitative determination of Calcium in Serum
For Use With (Equipment) Hitachi 747 Analyzers
Quantity 5 x 300 mL R2
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.