Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Calcium Reagents for Hitachi™ Analyzers
Shop All Pointe Scientific Products
Click to view available options
Quantity:
5 x 300 mL R2
5 x 600 mL R1
7 x 100 mL R1, 3 x 100 mL R2
7 x 50 mL R1, 3 x 50 mL R2
For Use With (Equipment):
Hitachi 704, 902 Analyzers
Hitachi 717, 911, 912, 914 Analyzers
Hitachi 747 Analyzers
Description
Includes
- Reagent 1: 2-Amino-2-Methyl-1-Propanol, pH 10.6±0.1
- Reagent 2: o-CPC 0.3mM, surfactant
- Method: Cresolphthalein Complexone (CPC)
- Format: Liquid (ready to use)
- Linearity: 20mg/dL
- Expected Values: 8.5 to 10.5mg/dL
Applications:
For quantitative determination of calcium in serum on Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Calcium Reagent Kit |
| Formulation | 2-Amino-2-Methyl-1-Propanol, pH 10.6 ±0.1 |
| For Use With (Application) | Quantitative determination of Calcium in Serum |
| For Use With (Equipment) | Hitachi 747 Analyzers |
| Quantity | 5 x 600 mL R1 |
| Form | Liquid |
For in vitro diagnostic use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction