Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific COBAS MIRA™ Reagent Containers and Sample Cups

Catalog No. 23666442 Shop All Pointe Scientific Products
Change view
Click to view available options
Volume (Metric):
4 mL
10 mL
Product Type:
Reagent Container
Sample Cup
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Type
23-666-442 10 mL Reagent Container
23-666-441 4 mL Reagent Container
23-666-440 - Sample Cup
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-442 Supplier Pointe Scientific Supplier No. 96MRC110

missing translation for 'validMainframeMsg'

Reagent containers and sample cups for use with the COBAS MIRA analyzer

Sizes for Reagent Containers: 4mL (0.14 oz.) and 10 mL (0.34 oz.)

Specifications

Product Type Reagent Container
Volume (Metric) 10 mL
For Use With (Equipment) Cobas Mira™ analyzer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.