Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Creatinine Reagents for Hitachi™ Analyzers

Catalog No. 23666233 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
11 x 100mL R1, 3 x 80mL R2
5 x 300 R2mL
5 x 600 R1mL
8 x 50mL R1, 2 x 50mL R2
Formulation:
Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer
Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer, Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-233 5 x 600 R1mL Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer
23-666-234 5 x 300 R2mL Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
23-666-232 8 x 50mL R1, 2 x 50mL R2 Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer, Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
23-666-231 11 x 100mL R1, 3 x 80mL R2 Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer, Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-666-233 Supplier Pointe Scientific Supplier No. HC439R1

Includes

  • Reagent 1: Alkaline buffer
  • Reagent 2: Picric Acid 40mM, surfactant

Quantify creatinine easily with system specific reagents

  • Method: Modified Jaffe, Initial Rate
  • Format: Liquid (ready to use)
  • Linearity: 25.0mg/dL
  • Expected Values: 0.40 to 1.40mg/dL

Specifications

Certifications/Compliance CE Marked
Type Creatinine R1
Formulation Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer
For Use With (Application) Quantitative determination of Creatinine in Serum
Quantity 5 x 600 R1mL
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.