Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific CRP (hs) Wide Range Reagent Sets

Catalog No. 23666462 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 20 mL R1, 1 x 20 mL R2
1 x 50 mL R1, 1 x 50 mL R2
5 x 2 mL
6 x 3 mL
Type:
CRP (hs) Calibrator Set
CRP (hs) Control Set
CRP (hs) Wide Range Reagent Set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-462 6 x 3 mL CRP (hs) Control Set
23-666-461 5 x 2 mL CRP (hs) Calibrator Set
23-666-460 1 x 50 mL R1, 1 x 50 mL R2 CRP (hs) Wide Range Reagent Set
23-666-459 1 x 20 mL R1, 1 x 20 mL R2 CRP (hs) Wide Range Reagent Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-462 Supplier Pointe Scientific Supplier No. C7568CTL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For quantitative determination of C-reactive protein in serum or plasma

  • Method: immunoturbidimetric
  • Latex particle enhanced assay
  • Reconstituted stability: ready-to-use liquid
  • Linearity: 0.1 to 320.0mg/L
  • Wavelength: 570nm
  • Expected values: <3mg/L

Specifications

Certifications/Compliance CE Marked
Type CRP (hs) Control Set
Formulation Level 1 CRP control, level 2 CRP control
For Use With (Application) Immunoturbidimetric assay
Quantity 6 x 3 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.