Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Detergent Reagents for Hitachi™ Analyzers

Catalog No. 23666252 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
14 x 100 mL
For Use With (Equipment):
Hitachi 911, 912, 914 analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Equipment)
23-666-252 14 x 100 mL Hitachi 911, 912, 914 analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-666-252 Supplier Pointe Scientific Supplier No. HI990DET

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Sodium hydroxide (1 to 5%); Ethanolamine (10 to 20%); EDTA (5 to 10%); Water (50 to 75%); Polyoxyethylene lauryl ether (1 to 5%); Emulgator K-30 (Paraffin oils and Kerosene) (1 to 5%)

Detergent solutions designed specifically for Hitachi Analyzers

  • Liquid format

For use with Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Detergent
For Use With (Equipment) Hitachi 911, 912, 914 analyzer
Quantity 14 x 100 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.