Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific G6PD Lyse Reagents

Catalog No. 23666474 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-474 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-474 Supplier Pointe Scientific Supplier No. G7583LYS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For quantitative, kinetic determination of Glucose-6-Phosphate deficiency in human red blood cells

  • Suitable for manual or automated use
  • Configuration: 1 × 100mL

Specifications

Certifications/Compliance CE Marked
Type G6PD Lyse Reagent
For Use With (Application) Quantitative determination of Fructosamine in Human Serum
Quantity 100 mL
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.