Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Glucose (Oxidase) Liquid Reagents

Catalog No. p-318806 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-286 1 x 120 mL
23-666-288 1 x 500 mL
23-666-287 1 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-286 Supplier Pointe Scientific Supplier No. G7521120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the quantitative determination of Glucose in serum. For in vitro diagnostic use only.

  • Reagent requires Glucose Standard (23-666-277)/Chemistry Calibrator (23-666-189)

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Type Glucose (Oxidase) Reagent Set
Formulation Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers
For Use With (Application) Quantitative determination of Glucose in Serum
Quantity 1 x 120 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.