Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | gamma-Glutamyl Transferase Reagent Set |
| Formulation | Tris Buffer 100mM, L-gamma-glutamyl-3-carboxy-4-nitroanilide >2.9mM, Glycylglycine 100mM, Sodium Azide 0.1%, Non-reactive Stabilizers and fillers |
| For Use With (Application) | Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum |
| Quantity | 10 x 10 mL |
| Form | Dry Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.