Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific γ-Glutamyl Transferase Reagents

Catalog No. 23666267 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
8 x 50 mL
10 x 10 mL
4 x 250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-267 8 x 50 mL
23-666-264 10 x 10 mL
23-666-265 4 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-267 Supplier Pointe Scientific Supplier No. G7570400

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance CE Marked
Type gamma-Glutamyl Transferase Reagent Set
Formulation Tris Buffer 100mM, L-gamma-glutamyl-3-carboxy-4-nitroanilide >2.9mM, Glycylglycine 100mM, Sodium Azide 0.1%, Non-reactive Stabilizers and fillers
For Use With (Application) Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum
Quantity 8 x 50 mL
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.