Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific HDL Cholesterol (PEG) Reagents

Catalog No. 23666301 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
2 x 30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-301 2 x 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-301 Supplier Pointe Scientific Supplier No. H751160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type HDL Cholesterol - PEG Reagent
Formulation HDL Cholesterol precipitating reagent: 20% w/v Polyethylene glycol in Glycine Buffer at pH 10 (25°C)
For Use With (Application) Quantitative determination of High Density Lipoprotein in Serum
Quantity 2 x 30 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.