Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific β-Hydroxybutyrate Reagents

Catalog No. 23666471 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 50 mL R1, 1 x 8 mL R2, 1 x 3 mL Std.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-471 1 x 50 mL R1, 1 x 8 mL R2, 1 x 3 mL Std.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-471 Supplier Pointe Scientific Supplier No. H758758
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Reagent 1: β-hydroxybutyrate dehydrogenase, diaphorase enzymes; Reagent 2: NAD, INT, oxalate; Reagent 3: Sodium D-3-hydroxybutyrate 1mM

  • Method: β-Hydroxybutyrate Dehydrogenase, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 505nm
  • Linearity: 4.5mM
  • Expected Values: 0.02 to 0.27mM

For the quantitative determination of β-hydroxybutyrate in serum or plasma.

Specifications

Certifications/Compliance CE Marked
Type beta-Hydroxybutyrate Reagent
Formulation R1 containing beta-hydroxybutyrate Dehydrogenase and Diaphorase Enzymes, R2 containing NAD, INT and Oxalate, Standard containing 1mM Sodium D-3-hydroxybutyrate
For Use With (Application) Determination of beta-hydroxybutyrate in Serum or Plasma
Quantity 1 x 50 mL R1, 1 x 8 mL R2, 1 x 3 mL Std.
Wavelength 505 nm
Form Liquid

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.