Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron/TIBC Reagents

Catalog No. 23666319 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
400 Tests
60 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-319 400 Tests
23-666-320 60 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-319 Supplier Pointe Scientific Supplier No. I7504400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Iron/TIBC Reagent Kit
Formulation Iron Buffer contains: Acetic Acid (dilute) 6%, UIBC Buffer contains: Sodium Azide 0.05%, Iron color contains: Ferrozine 16.7mM, Iron standard contains: Ferrous Chloride
For Use With (Application) Quantitative determination of Iron and total Iron-binding capacity in Serum
Includes 1 x 1L Iron Buffer, 1 x 1L UIBC Buffer, 1 x 60mL Iron Color, 1 x 500mL Iron Standard
Quantity 400 Tests
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.