Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron-Total Liquid Reagents

Catalog No. 23666318 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL Buffer, 1 x 5 mL Color
1 x 500 mL Buffer, 1 x 15 mL Color
5 x 50mL R1, 1 x 50mL R2
Type:
Iron (Ferrozine) Reagent
Total Iron Reagent Set 1
Total Iron Reagent Set 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-318 1 x 500 mL Buffer, 1 x 15 mL Color Total Iron Reagent Set 2
23-666-317 1 x 120 mL Buffer, 1 x 5 mL Color Total Iron Reagent Set 1
23-666-316 5 x 50mL R1, 1 x 50mL R2 Iron (Ferrozine) Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-318 Supplier Pointe Scientific Supplier No. I7505500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5); surfactant
  • Color Reagent: Ferrozine 16.7mM in hydroxylamine hydrochloride

Quantify iron levels in serum. Use with Pointe Scientific™ Iron Standard Reagent (Catalog No. 23-666-312)

  • Method: Ferrozine, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 560nm
  • Linearity: 500ug/dL
  • Expected Values: 60 to 150ug/dL

For the quantitative determination of iron in serum

Specifications

Certifications/Compliance CE Marked
Type Total Iron Reagent Set 2
Formulation Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, Iron color R2: 3.6mM Ferrozine in Hydroxylamine Hydrochloride
For Use With (Application) Quantitative determination of Iron in Serum
Quantity 1 x 500 mL Buffer, 1 x 15 mL Color
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.