Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron-Total Reagents for Hitachi Analyzers

Catalog No. 23666315 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
3 x 500 mL R1
5 x 200 R2mL
Formulation:
Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer pH 4.5 with Surfactant
Iron color R2: Ferrozine 3.6mM in Hydroxylamine Hydrochloride
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-315 5 x 200 R2mL Iron color R2: Ferrozine 3.6mM in Hydroxylamine Hydrochloride
23-666-314 3 x 500 mL R1 Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer pH 4.5 with Surfactant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-666-315 Supplier Pointe Scientific Supplier No. HI404R2

missing translation for 'validMainframeMsg'

Includes

  • Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5) with surfactant
  • Color Reagent: Ferrozine 3.6mM in hydroxylamine hydrochloride

Requires

Requires calibrator/standard

Quantify iron levels in serum

  • Method: Ferrozine
  • Format: Liquid (ready to use)
  • Linearity: 500ug/dL
  • Expected Values: 60 to 150ug/dL

Applications:

For the quantitative determination of iron in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Total Iron Reagent Kit
Formulation Iron color R2: Ferrozine 3.6mM in Hydroxylamine Hydrochloride
For Use With (Application) Quantitative determination of Iron in Serum
Quantity 5 x 200 R2mL
Form Liquid

For in vitro diagnostics use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.