Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Iron-UIBC/TIBC Buffer Reagent

Catalog No. 23666423 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-423 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-423 Supplier Pointe Scientific Supplier No. I7506500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Iron/UIBC/TIBC Reagent
Formulation Iron Buffer reagent: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, UIBC Buffer reagent: Tris 500mM, pH 8.1 with Surfactant, Sodium Azide as preservative, Iron color reagent: 16.7mM Ferrozine in Hydroxylamine Hydrochloride, Iron standard Ferrous Chloride in Hydroxylamine Hydrochloride
For Use With (Application) Quantitative determination of Iron and total Iron-binding capacity in Serum
Quantity 1 x 500 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.