Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE Diluent

Catalog No. 23666329 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 300 mL
6 x 1 L
6 x 2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-329 5 x 300 mL
23-666-328 6 x 2 L
23-666-326 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-329 Supplier Pointe Scientific Supplier No. HI990DIL

missing translation for 'validMainframeMsg'

Includes

Borate buffer with anti-microbial (0.05%)

For sodium, potassium and chloride analysis with Hitachi™ and Roch™e systems

  • Method: Ion-Selective Electrode (ISE)
  • Format: Liquid

ISE Diluent, ISE Internal Reference and ISE 1N KCl are for the quantitative determination of sodium, potassium, and chloride in serum by ion-selective electrode methods on Roche/Hitachi systems

Specifications

Certifications/Compliance CE Marked
Type ISE Diluent
Formulation Borate Buffer 0.05% nonreactive ingredient Anti-microbial
For Use With (Application) Quantitative determination of Sodium, Potassium and Chloride in Serum
Quantity 5 x 300 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.