Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE Internal Reference Reagents

Catalog No. 23666332 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 600 mL
6 x 2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-332 5 x 600 mL
23-666-321 6 x 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-332 Supplier Pointe Scientific Supplier No. HI990REF

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Borate buffer: Sodium chloride 3.2mM; Sodium bicarbonate 1.3mM; Potassium phosphate 0.15mM; anti-microbial (0.05%)

Used for sodium, potassium and chloride analysis with Hitachi™ and Roche™ systems

  • Method: Ion-Selective Electrode (ISE)
  • Format: Liquid

ISE Diluent, ISE Internal Reference and ISE 1N KCl are for the quantitative determination of sodium, potassium, and chloride in serum by ion-selective electrode methods on Roche/Hitachi systems

Specifications

Certifications/Compliance CE Marked
Type ISE Internal Reference Solution
Formulation ISE Internal Reference: Borate Buffer- Sodium Chloride 3.2mM/L, Sodium Bicarbonate 1.3mM/L, Potassium Phosphate 0.15mM/L, Anti-microbial Nonreactive Ingredient 0.05%
For Use With (Application) Quantitative determination of Sodium, Potassium and Chloride in Serum
Quantity 5 x 600 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.