Learn More
Pointe Scientific ISE Solution Reagents for Roche Cobas™ Analyzers
Shop All Pointe Scientific ProductsDescription
- Method: Ion-selective Electrode (ISE)
- Format: Liquid
- Reference Solution contains: KCl 3.5mM
- Standard Solution 1 contains: Na+ 150mM; K+ 3mM; Cl- 138mM; Preservative
- Standard Solution 2 contains: Na+ 100mM; K+ 7mM; Cl- 92mM; Preservative
- Etcher Solution contains: Ammonium hydrogen difluoride 0.6%
For ISE applications on Roche Cobas Chemistry Systems
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | ISE Solution Reagent |
| Formulation | KCl aqueous solution 3.5mM/L |
| Quantity | 60 mL |
| Form | Liquid |
For in vitro diagnostic use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.