Learn More
Description
- Method: Ion-Specific Electrode (ISE)
- Format: Liquid (ready to use)
- Configuration: 10 ampules × 2.0mL
- ISE Standard Low contains: Na+ 120.0mM; K+ 3.0mM; Cl- 80.0mM
- ISE Standard High contains: Na+ 160.0mM; K+ 7.0mM; Cl- 120.0mM
- For the standardization of Sodium, Potassium and Chloride on Ion-Selective Electrodes
- Intended for use with Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | ISE Standard High Reagent Kit |
| Formulation | NA+ 160mM/L, K+ 7mM/L, Cl- 120mM/L |
| For Use With (Application) | Standardization of Sodium, Potassium and Chloride on ion selective electrodes |
| Quantity | 10 x 2 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.