Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Lactate Dehydrogenase Dry Powder Reagents

Catalog No. 23666353 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-353 4 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-666-353 Supplier Pointe Scientific Supplier No. L75351L

missing translation for 'validMainframeMsg'

Includes

NAD 7.0 mM; L-Lactate 50mM; Tris Buffer (pH 9.0±0.1); Nonreactive stabilizers and fillers

Quantify lactate dehydrogenase activity in serum

  • Method: Modified Wacker/Tris, Kinetic
  • Format: Dry powder
  • Wavelength: 340nm
  • Linearity: 800 IU/L
  • Expected Values: 80 to 285 U/L (Male, 37°C); 103 to 227 U/L (Female, 37°C)
  • Reconstituted stability: 8 hours RT, 21 days at 2° to 8°C

For the in vitro quantitative kinetic determination of lactate dehydrogenase activity in serum

Specifications

Certifications/Compliance CE Marked
Type Lactate Dehydrogenase Reagent Set
Formulation NAD 7mM, L-Lactate 50mM, Tris Buffer pH 9 ± 0.1, non-reactive Stabilizers and fillers
For Use With (Application) Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum, in-vitro diagnostic use only
pH 9
Quantity 4 x 250 mL
Wavelength 340 nm
Form Dry Powder

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.