Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Lactate Dehydrogenase Liquid Reagents

Catalog No. 23666348 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120mL R1, 1 x 30mL R2
1 x 500mL R1, 1 x 125mL R2
3 x 120mL R1, 1 x 90mL R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-348 1 x 120mL R1, 1 x 30mL R2
23-666-350 1 x 500mL R1, 1 x 125mL R2
23-666-349 3 x 120mL R1, 1 x 90mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-348 Supplier Pointe Scientific Supplier No. L7572150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Buffer and Co-Enzyme when combined: NAD 5.8mM; L-Lactate 55mM; Buffer (pH 8.95); Non-reactive stabilizers; Sodium azide 0.1%

Quantify lactate dehydrogenase activity in serum

  • Method: Modified Wacker/Tris, Kinetic
  • Format: Liquid (ready to use)
  • Wavelength: 340nm
  • Linearity: 1000 U/L
  • Expected Values: 80 to 285 U/L (Male, 37°C); 103 to 227 U/L (Female, 37°C)

For the in vitro quantitative kinetic determination of lactate dehydrogenase activity in serum

Specifications

Certifications/Compliance CE Marked
Type Lactate Dehydrogenase Reagent Kit
Formulation NAD 5.8mM, L-Lactate 55mM, 8.95 Buffer pH, non-reactive Stabilizers and Sodium Azide 0.1% as preservative
For Use With (Application) Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum
Quantity 1 x 120mL R1, 1 x 30mL R2
Wavelength 340 nm
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.